4-Iodo-2-nitrophenol 95%

Supplier: Ambeed
Danger

A199469-1G A199469-250MG A199469-5G A199469-25G
76986-390EA 14 USD
76986-390 76986-392 76986-394 77567-868
4-Iodo-2-nitrophenol 95%
4-Iodo-2-nitrophenol

Precaución: For research use only.

Formula: C₆H₄INO₃
MDL Number: MFCD00466297
CAS Number: 21784-73-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR