4-Methoxycarbonyl-3-nitrophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Methoxycarbonyl)-3-nitrophenyl)boronic acid

A199416-1G A199416-250MG A199416-5G
77030-086EA 32 USD
77030-086 77030-088 77030-090
4-Methoxycarbonyl-3-nitrophenylboronic acid 98%
4-Methoxycarbonyl-3-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₆
MW: 224.963 g/mol
MDL Number: MFCD12032027
CAS Number: 85107-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR