1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) 95%

Supplier: Ambeed
Warning

Synonyms: 1,4-Cyclohexanedicarboxylic acid

A199359-100G A199359-25G A199359-500G
76999-268EA 23 USD
76999-268 76999-270 76999-272
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) 95%
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers)

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
MDL Number: MFCD00001465
CAS Number: 1076-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR