3-Pyridineboronic acid hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Pyridin-3-ylboronic acid hydrochloride

A199120-1G A199120-250MG A199120-5G
76999-256EA 6 USD
76999-256 76999-258 76999-260
3-Pyridineboronic acid hydrochloride 95%
3-Pyridineboronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₇BClNO₂
MDL Number: MFCD04112572
CAS Number: 265664-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR