Methyl (2-chloromethyl)oxazole-4-carboxylate 98%
Supplier: Ambeed
|
Danger
|
A198948-10G
A198948-1G
A198948-250MG
A198948-5G
77012-586EA
303
USD
77012-586
77012-588
77012-590
77890-202
Methyl (2-chloromethyl)oxazole-4-carboxylate 98%
Methyl (2-chloromethyl)oxazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₆ClNO₃ |
MDL Number:
MFCD06660133 CAS Number: 208465-72-9 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...