2-Chloro-6-fluoro-3-methylphenylboronic acid 95%

Supplier: Ambeed
Warning

A198476-1G A198476-250MG A198476-5G
77030-064EA 16 USD
77030-064 77030-066 77030-068
2-Chloro-6-fluoro-3-methylphenylboronic acid 95%
2-Chloro-6-fluoro-3-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BClFO₂
MDL Number: MFCD03701534
CAS Number: 352535-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR