GDC-0941 98+%

Supplier: Ambeed
Warning

A198456-10MG A198456-1MG A198456-50MG A198456-5MG
77030-056EA 29.71 USD
77030-056 77030-058 77030-060 77030-062
GDC-0941 98+%
GDC-0941

Precaución: for Research Use Only

Formula: C₂₃H₂₇N₇O₃S₂
MW: 513.6 g/mol
MDL Number: MFCD11616196
CAS Number: 957054-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR