4'-Hydroxyvalerophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Hydroxyphenyl)-1-pentanone

A198418-100G A198418-25G A198418-5G
76999-228EA 40.32 USD
76999-228 76999-230 76999-232
4'-Hydroxyvalerophenone 97%
4'-Hydroxyvalerophenone

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00009719
CAS Number: 2589-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR