5-Bromo-2-propoxypyridine 98%

Supplier: Ambeed
Warning

A198254-1G A198254-250MG A198254-5G
77030-050EA 94.43 USD
77030-050 77030-052 77030-054
5-Bromo-2-propoxypyridine 98%
5-Bromo-2-propoxypyridine

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MW: 216.07 g/mol
MDL Number: MFCD09743641
CAS Number: 850142-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR