6-Bromo-3-methyl-3H-imidazo[4,5-b]pyridine 98%

Supplier: Ambeed
Warning

A198065-1G A198065-250MG A198065-5G A198065-25G
76999-218EA 97.61 USD
76999-218 76999-220 76999-222 77671-752
6-Bromo-3-methyl-3H-imidazo[4,5-b]pyridine 98%
6-Bromo-3-methyl-3H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MDL Number: MFCD08692133
CAS Number: 37805-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR