(1S,4S)-Dimethylcyclohexane-1,4-dicarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: cis-Dimethyl cyclohexane-1,4-dicarboxylate

A198054-100MG A198054-1G A198054-250MG A198054-5G
76986-354EA 16.98 USD
76986-354 76986-356 76986-358 76986-360
(1S,4S)-Dimethylcyclohexane-1,4-dicarboxylate 95%
(1S,4S)-Dimethylcyclohexane-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₄
MW: 200.24 g/mol
MDL Number: MFCD00216473
CAS Number: 3399-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR