tert-Butyl ((3S,5S)-5-(hydroxymethyl)pyrrolidin-3-yl)carbamate 98%
Supplier: Ambeed
|
Warning
|
A197791-100MG
A197791-250MG
77012-554EA
22
USD
77012-554
77012-556
tert-Butyl ((3S,5S)-5-(hydroxymethyl)pyrrolidin-3-yl)carbamate 98%
tert-Butyl ((3S,5S)-5-(hydroxymethyl)pyrrolidin-3-yl)carbamate
Precaución: for Research Use Only
Formula:
C₁₀H₂₀N₂O₃ |
MDL Number:
MFCD08704519 CAS Number: 663948-85-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...