3-Amino-5-chloro-2-hydroxypyridine 97%

Supplier: Ambeed
Warning

A197781-250MG A197781-5G
76986-348EA 12 USD
76986-348 76986-350
3-Amino-5-chloro-2-hydroxypyridine 97%
3-Amino-5-chloro-2-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MW: 144.55 g/mol
MDL Number: MFCD07375031
CAS Number: 98027-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR