5,6-Dichloro-1H-indazole 97%

Supplier: Ambeed
Warning

A197620-100MG A197620-1G A197620-250MG
76986-342EA 31.83 USD
76986-342 76986-344 76986-346
5,6-Dichloro-1H-indazole 97%
5,6-Dichloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂
MW: 187.0300 g/mol
MDL Number: MFCD11878111
CAS Number: 124691-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR