2-(Benzyloxy)-6-chloropyridin-4-amine 95%

Supplier: Ambeed
Warning

A197519-1G A197519-250MG
76986-334EA 476.35 USD
76986-334 76986-336
2-(Benzyloxy)-6-chloropyridin-4-amine 95%
2-(Benzyloxy)-6-chloropyridin-4-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₁ClN₂O
MW: 234.68 g/mol
MDL Number: MFCD20257883
CAS Number: 1346809-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR