Ethyl 3-(Difluoromethyl)-1-methylpyrazole-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carboxylate, 3-(Difluoromethyl)-1-methylpyrazole-4-carboxylic acid Ethyl ester

A197104-10G A197104-1G A197104-25G A197104-5G
76986-326EA 98.67 USD
76986-326 76986-328 76986-330 76986-332
Ethyl 3-(Difluoromethyl)-1-methylpyrazole-4-carboxylate 97%
Ethyl 3-(Difluoromethyl)-1-methylpyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₀F₂N₂O₂
MW: 204.18 g/mol
MDL Number: MFCD12022341
CAS Number: 141573-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR