cis-Ethyl 3-aminocyclobutanecarboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A196768-1G A196768-250MG A196768-5G
76986-320EA 200 USD
76986-320 76986-322 76986-324
cis-Ethyl 3-aminocyclobutanecarboxylate hydrochloride 97%
cis-Ethyl 3-aminocyclobutanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₂
MDL Number: MFCD12925784
CAS Number: 957793-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR