3-Iodo-2-methoxyisonicotinonitrile 95%

Supplier: Ambeed
Warning

A196526-1G A196526-250MG
76986-306EA 142.17 USD
76986-306 76986-308
3-Iodo-2-methoxyisonicotinonitrile 95%
3-Iodo-2-methoxyisonicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₅IN₂O
MW: 260.03 g/mol
MDL Number: MFCD09056806
CAS Number: 908279-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR