7-Bromo-5-nitro-1H-indazole 96%

Supplier: Ambeed
Warning

A196515-100MG A196515-10G A196515-1G A196515-250MG A196515-25G A196515-5G
76999-170EA 15.92 USD
76999-170 76999-172 76999-174 76999-176 76999-178 76999-180
7-Bromo-5-nitro-1H-indazole 96%
7-Bromo-5-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MDL Number: MFCD03617603
CAS Number: 685109-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR