Ethyl-4-hydroxy-8-methylquinoline-3-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 8-methyl-4-oxo-1,4-dihydroquinoline-3-carboxylate
A196439-10G
A196439-1G
A196439-250MG
A196439-25G
A196439-5G
77029-996EA
183.54
USD
77029-996
77029-998
77030-000
77030-002
77030-004
Ethyl-4-hydroxy-8-methylquinoline-3-carboxylate 95%
Ethyl 4-hydroxy-8-methylquinoline-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₃H₁₃NO₃ MW: 231.2500 g/mol |
MDL Number:
MFCD00173390 CAS Number: 77156-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...