5-Methoxy-1H-pyrrolo[3,2-b]pyridine 98%

Supplier: Ambeed
Danger

A196381-1G A196381-250MG A196381-5G
77012-520EA 53.05 USD
77012-520 77012-522 77611-892
5-Methoxy-1H-pyrrolo[3,2-b]pyridine 98%
5-Methoxy-1H-pyrrolo[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C8H8N2O
MW: 148.1619 g/mol
MDL Number: MFCD08688601
CAS Number: 17288-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR