3-Methyl-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: H-Phe(3-Me)-OH

A196370-250MG A196370-1G
76999-150EA 21.22 USD
76999-150 77600-600
3-Methyl-L-phenylalanine 97%
3-Methyl-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD00044972
CAS Number: 114926-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR