2-(4-Fluorophenoxy)ethanamine hydrochloride 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A196354-250MG A196354-5G
76986-298EA 8.49 USD
76986-298 77683-536
2-(4-Fluorophenoxy)ethanamine hydrochloride 97%, Ambeed.Inc
2-(4-Fluorophenoxy)ethanamine hydrochloride

Precaución: For research use only.

Formula: C₈H₁₁ClFNO
MW: 191.63 g/mol
MDL Number: MFCD09971613
CAS Number: 263409-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR