trans-N1,N1-Dimethylcyclohexane-1,2-diamine 95%

Supplier: Ambeed
Warning

A196350-10G A196350-1G A196350-25G
77012-514EA 44 USD
77012-514 77012-516 77012-518
trans-N1,N1-Dimethylcyclohexane-1,2-diamine 95%
trans-N1,N1-Dimethylcyclohexane-1,2-diamine

Precaución: for Research Use Only

Formula: C₈H₁₈N₂
MDL Number: MFCD09838678
CAS Number: 67198-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR