(4-Bromobenzyl)triphenylphosphonium bromide 97%

Supplier: Ambeed
Warning

A196180-100G A196180-10G A196180-25G A196180-5G
77029-986EA 80 USD
77029-986 77029-988 77029-990 77029-992
(4-Bromobenzyl)triphenylphosphonium bromide 97%
(4-Bromobenzyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₅H₂₁Br₂P
MDL Number: MFCD00051922
CAS Number: 51044-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR