3-Amino-6-(trifluoromethoxy)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 6-(Trifluoromethoxy)pyridin-3-amine

A195888-1G A195888-250MG
77012-500EA 1400.39 USD
77012-500 77012-502
3-Amino-6-(trifluoromethoxy)pyridine 97%
3-Amino-6-(trifluoromethoxy)pyridine

Precaución: for Research Use Only

Formula: C₆H₅F₃N₂O
MW: 178.112 g/mol
MDL Number: MFCD07374879
CAS Number: 135900-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR