5-Bromo-4-fluoro-2-methoxybenzoic acid 97%

Supplier: Ambeed
Warning

A195839-100MG A195839-250MG A195839-5G
76986-286EA 21.22 USD
76986-286 76986-288 76986-290
5-Bromo-4-fluoro-2-methoxybenzoic acid 97%
5-Bromo-4-fluoro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₃
MW: 249.034 g/mol
MDL Number: MFCD15527529
CAS Number: 95383-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR