2-Methylisoquinolin-1(2H)-one 98%
Supplier: Ambeed
|
Warning
|
A195833-10G
A195833-1G
A195833-250MG
A195833-25G
A195833-500G
A195833-5G
76986-274EA
246
USD
76986-274
76986-276
76986-278
76986-280
76986-282
76986-284
2-Methylisoquinolin-1(2H)-one 98%
2-Methylisoquinolin-1(2H)-one
Precaución: for Research Use Only
Formula:
C₁₀H₉NO |
MDL Number:
MFCD00040275 CAS Number: 4594-71-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...