7-(Trifluoromethyl)chroman-4-one 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydro-7-(trifluoromethyl)-4H-chromen-4-one

A195809-250MG A195809-1G A195809-5G
77029-962EA 24 USD
77029-962 77600-594 77871-978
7-(Trifluoromethyl)chroman-4-one 97%
7-(Trifluoromethyl)chroman-4-one

Precaución: for Research Use Only

Formula: C₁₀H₇F₃O₂
MW: 216.157 g/mol
MDL Number: MFCD09998165
CAS Number: 111141-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR