1-Hydroxycyclohexyl phenyl ketone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1-Hydroxycyclohexyl)(phenyl)methanone
A195514-1000G
A195514-100G
A195514-500G
A195514-25G
77029-950EA
27
USD
77029-950
77029-952
77029-954
77029-956
77029-958
77029-960
77085-324
1-Hydroxycyclohexyl phenyl ketone 98%
1-Hydroxycyclohexyl phenyl ketone
Precaución: for Research Use Only
Formula:
C₁₃H₁₆O₂ |
MDL Number:
MFCD00059561 CAS Number: 947-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...