1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19-hexaoxa-4-azadocosan-22-oic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-amino-PEG5-acid
A195206-100MG
A195206-1G
A195206-250MG
A195206-25G
A195206-5G
76986-248EA
31.83
USD
76986-248
76986-250
76986-252
77683-528
77683-529
1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19-hexaoxa-4-azadocosan-22-oic acid 95%
1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19-hexaoxa-4-azadocosan-22-oic acid
Precaución: For research use only.
Formula:
C₂₈H₃₇NO₉ MW: 531.59 g/mol |
MDL Number:
MFCD06656473 CAS Number: 882847-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...