1-Ethyl-1H-pyrazol-5-ylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (1-Ethyl-1H-pyrazol-5-yl)boronic acid

A195080-100MG A195080-250MG A195080-1G A195080-5G
76986-244EA 22.28 USD
76986-244 76986-246 77611-874 77611-876
1-Ethyl-1H-pyrazol-5-ylboronic acid 97%
1-Ethyl-1H-pyrazol-5-ylboronic acid

Precaución: for Research Use Only

Formula: C₅H₉BN₂O₂
MW: 139.948 g/mol
MDL Number: MFCD03419491
CAS Number: 1095080-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR