N-Methoxy-N-methyl-4-(trifluoromethyl)benzenecarboxamide 98%
Supplier: Ambeed
|
Warning
|
A194931-1G
A194931-25G
A194931-5G
76999-098EA
13
USD
76999-098
76999-100
76999-102
N-Methoxy-N-methyl-4-(trifluoromethyl)benzenecarboxamide 98%
N-Methoxy-N-methyl-4-(trifluoromethyl)benzenecarboxamide
Precaución: for Research Use Only
Formula:
C₁₀H₁₀F₃NO₂ |
MDL Number:
MFCD08689795 CAS Number: 116332-61-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...