2-Amino-5-bromothiazole-4-carbonitrile 95%

Supplier: Ambeed
Warning

A194785-100MG A194785-250MG A194785-1G
76986-234EA 50.93 USD
76986-234 76986-236 77683-523
2-Amino-5-bromothiazole-4-carbonitrile 95%
2-Amino-5-bromothiazole-4-carbonitrile

Precaución: For research use only.

Formula: C₄H₂BrN₃S
MW: 204.05 g/mol
MDL Number: MFCD11109851
CAS Number: 944804-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR