2-Methylnicotinonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 3-Cyano-2-methylpyridine

A194772-100MG A194772-1G A194772-250MG
77012-472EA 31.83 USD
77012-472 77012-474 77012-476
2-Methylnicotinonitrile 95%
2-Methylnicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₆N₂
MW: 118.14 g/mol
MDL Number: MFCD06254420
CAS Number: 1721-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR