3-Amino-5,6,7,8-tetrahydro-6-methyl-1,6-naphthyridine 95%

Supplier: Ambeed
Warning

A194771-100MG A194771-1G A194771-250MG
76999-092EA 87 USD
76999-092 76999-094 76999-096
3-Amino-5,6,7,8-tetrahydro-6-methyl-1,6-naphthyridine 95%
3-Amino-5,6,7,8-tetrahydro-6-methyl-1,6-naphthyridine

Precaución: for Research Use Only

Formula: C9H13N3
MW: 163.2196 g/mol
MDL Number: MFCD09743968
CAS Number: 216966-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR