(2,5-Dibromophenyl)hydrazine 97%

Supplier: Ambeed
Warning

A194769-1G A194769-250MG
76986-230EA 63.66 USD
76986-230 76986-232
(2,5-Dibromophenyl)hydrazine 97%
(2,5-Dibromophenyl)hydrazine

Precaución: for Research Use Only

Formula: C₆H₆Br₂N₂
MW: 265.933 g/mol
MDL Number: MFCD02663135
CAS Number: 62672-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR