6,8-Dichloroimidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A194734-1G A194734-5G
76999-088EA 18.04 USD
76999-088 76999-090
6,8-Dichloroimidazo[1,2-a]pyridine 97%
6,8-Dichloroimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂
MW: 187.03 g/mol
MDL Number: MFCD06761692
CAS Number: 858516-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR