4-Chloro-3-(trifluoromethoxy)bromobenzene 98%

Supplier: Ambeed
Warning

A194534-1G A194534-5G A194534-25G
77012-456EA 17 USD
77012-456 77012-458 77892-257
4-Chloro-3-(trifluoromethoxy)bromobenzene 98%
4-Chloro-3-(trifluoromethoxy)bromobenzene

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃O
MDL Number: MFCD09878140
CAS Number: 406232-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR