2-Bromo-3-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A194314-1G A194314-250MG A194314-5G
77029-920EA 27 USD
77029-920 77029-922 77871-967
2-Bromo-3-(trifluoromethyl)benzoic acid 97%
2-Bromo-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₂
MDL Number: MFCD08532493
CAS Number: 177420-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR