4-Amino-5-chloro-2-ethoxy-N-((4-(4-fluorobenzyl)-2-morpholinyl)methyl)benzamide ≥99%

Supplier: Ambeed
Warning

Synonyms: Mosapride, 4-Amino-5-chloro-2-ethoxy-N-((4-(4-fluorobenzyl)morpholin-2-yl)methyl)benzamide

A194108-10G A194108-10MG A194108-50MG
76999-082EA 105.03 USD
76999-082 76999-084 76999-086
4-Amino-5-chloro-2-ethoxy-N-((4-(4-fluorobenzyl)-2-morpholinyl)methyl)benzamide ≥99%
4-Amino-5-chloro-2-ethoxy-N-((4-(4-fluorobenzyl)-2-morpholinyl)methyl)benzamide

Precaución: for Research Use Only

Formula: C₂₁H₂₅ClFN₃O₃
MW: 421.89 g/mol
MDL Number: MFCD00867430
CAS Number: 112885-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR