N-(4-Methoxybenzyl)-1-(naphthalen-1-yl)methanamine 97%

Supplier: Ambeed
Warning

A193954-100MG A193954-250MG
76986-204EA 41.38 USD
76986-204 76986-206
N-(4-Methoxybenzyl)-1-(naphthalen-1-yl)methanamine 97%
N-(4-Methoxybenzyl)-1-(naphthalen-1-yl)methanamine

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO
MW: 277.36 g/mol
MDL Number: MFCD01469095
CAS Number: 185669-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR