Ethyl 2-bromopyrimidine-4-carboxylate 95%

Supplier: Ambeed
Warning

A193870-100MG A193870-1G A193870-250MG
77012-450EA 98.67 USD
77012-450 77012-452 77012-454
Ethyl 2-bromopyrimidine-4-carboxylate 95%
Ethyl 2-bromopyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MW: 231.04 g/mol
MDL Number: MFCD27923790
CAS Number: 1207954-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR