Methyl non-2-enoate 96%

Supplier: Ambeed
Warning

A193869-100G A193869-10G A193869-5G A193869-25G
76999-076EA 54.11 USD
76999-076 76999-078 76999-080 77302-370
Methyl non-2-enoate 96%
Methyl non-2-enoate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
MDL Number: MFCD00027316
CAS Number: 111-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR