4-Nitrobenzonitrile 97%

Supplier: Ambeed
Danger

A193835-100G A193835-5G A193835-25G
77029-914EA 93 USD
77029-914 77029-916 77085-100
4-Nitrobenzonitrile 97%
4-Nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄N₂O₂
MDL Number: MFCD00007279
CAS Number: 619-72-7
UN: 3439
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR