tert-Butyl (2-bromothiazol-5-yl)carbamate 97%

Supplier: Ambeed
Warning

A193722-100MG A193722-1G A193722-250MG A193722-5G
77029-902EA 14 USD
77029-902 77029-904 77029-906 77029-908 77029-910
tert-Butyl (2-bromothiazol-5-yl)carbamate 97%
tert-Butyl (2-bromothiazol-5-yl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₂O₂S
MDL Number: MFCD13195442
CAS Number: 1094070-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR