1-(2,6-Diisopropylphenyl)-2-phenyl-1H-imidazole 95%

Supplier: Ambeed
Warning

A193706-100MG A193706-1G A193706-250MG
77012-444EA 35.01 USD
77012-444 77012-446 77012-448
1-(2,6-Diisopropylphenyl)-2-phenyl-1H-imidazole 95%
1-(2,6-Diisopropylphenyl)-2-phenyl-1H-imidazole

Precaución: for Research Use Only

Formula: C₂₁H₂₄N₂
MW: 304.43 g/mol
MDL Number: MFCD27923076
CAS Number: 914306-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR