6-Chloro-2-morpholinopyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A193633-250MG A193633-5G A193633-1G
77012-440EA 14 USD
77012-440 77012-442 77890-185
6-Chloro-2-morpholinopyrimidin-4-amine 95%
6-Chloro-2-morpholinopyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₈H₁₁ClN₄O
MDL Number: MFCD09864565
CAS Number: 3549-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR