8-Azaspiro[4.5]decane hydrochloride 98%

Supplier: Ambeed
Warning

A193433-1G A193433-250MG A193433-5G
76986-188EA 117.76 USD
76986-188 76986-190 77683-517
8-Azaspiro[4.5]decane hydrochloride 98%
8-Azaspiro[4.5]decane hydrochloride

Precaución: For research use only.

Formula: C₉H₁₈ClN
MW: 175.69 g/mol
MDL Number: MFCD09759059
CAS Number: 1123-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR