2-(4-Amino-1H-pyrazol-1-yl)ethanol 98%

Supplier: Ambeed
Warning

A193321-100MG A193321-1G A193321-250MG A193321-5G
77029-890EA 10 USD
77029-890 77029-892 77029-894 77029-896 77029-898
2-(4-Amino-1H-pyrazol-1-yl)ethanol 98%
2-(4-Amino-1H-pyrazol-1-yl)ethanol

Precaución: for Research Use Only

Formula: C₅H₉N₃O
MDL Number: MFCD11147853
CAS Number: 948571-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR